missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Justrite™ Containers
VapLock™ Solvent Waste Manifold Caps for Justrite™ Containers connect to waste containers for a secure closed system
Specifications
For Use With (Application) | Solvent Safety |
---|---|
For Use With (Equipment) | Solvent-Compatible Containers |
Description
Different materials, numbers and sizes of ports help customize to your applicationWarranty
None
Specifications
Solvent Safety | |
Solvent-Compatible Containers |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title