missing translation for 'onlineSavingsMsg'
Learn More

Welch Ilmvac Vacuum controller 195mm x 178mm x 105mm with 8mm hose tail connections and ceramic sensor

Brand:  Welch Ilmvac VCB424

Vacuum controller 195mm x 178mm x 105mm with 8mm hose tail connections and ceramic sensor

Code : 04

Additional Details : Weight : 1.00000kg

Product Code. 12975154

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers