Learn More
Memmert™ Usb User-Id Stick (With User-Id Licence), For Uf160Ts
Memmert™ USB User-ID Stick prevents undesired manipulation by unauthorized third parties.
Brand: Memmert™ UF160TSB33170
Includes: User-ID License
Description
- USB User-ID stick (with User-ID licence)
- Oven-linked authorisation licence (User-ID-programme) on Memory-stick
- Prevents undesired manipulation by unauthorised third parties
- When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
Only for units with TwinDISPLAY | |
Units With TwinDISPLAY | |
USB Stick |