missing translation for 'onlineSavingsMsg'
Learn More

Ubiquitin Rabbit Polyclonal Antibody, Epredia™

Ubiquitin, Rabbit Polyclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Brand:  Epredia RB-9203-P1

Code : LD

Additional Details : Weight : 0.00190kg

 View more versions of this product

Product Code. 12683467

  • £355.00 / 500µL
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ubiquitin (Ub) plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The protein to be degraded is conjugated to Ub and the ubiquinated protein is then selectively degraded by a 26S complex, multicatalytic cytosolic and nuclear protease termed proteasome. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Recombinant full-length human ubiquitin protein

Molecular Weight: 8.5kDa

Positive Control: Human brain from patient with Alzheimer's disease

Cellular Localization: Nuclear, cytoplasmic and membrane This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Ubiquitin
Polyclonal
Unconjugated
Recombinant full-length human ubiquitin protein
IVD
Human
IgG
Immunohistochemistry (Paraffin)
200µg/mL
Rabbit
500 μL
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers