Learn More
DuPont™ Tyvek™ 500 Laboratory Coats Press Studs Product Code.: 15541362
DuPont™ Tyvek™ 500 Laboratory Coats Press Studs
Product Code. 15541362 Shop All DuPont Products
Medium
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Product Code. 15541362
Brand: DuPont™ D13396079
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Offers enhanced protection for body parts that are more exposed to hazardous substances, fine particles, and fibres. Other features include ultra low-linting and antistatically treated.
  • Silicon non-added
  • Stitched internal seams
  • 3 pockets and 5 press stud closures
  • Composed of flash spun high density polyethylene
  • Provides an idea balance of protection, durability and comfort
  • Permeable to both air and water vapor, yet repels water-based liquids and aerosols
  • Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Resistant Lab Coat
Size Medium
Gender Unisex
Material HDPE
Closure Type Press Studs
Disposable Limited-use
Sterility Non-sterile
Resistance Chemicals
Product Content Correction
Product Title
DuPont™ Tyvek™ 500 Laboratory Coats Press Studs >