missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX Vol.2-90 Right-Hinged
Safe and approved storage of flammable hazardous substances in working areas
£2541.88 - £3342.18
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Material | Steel |
Product Type | Safety storage cabinet |
Door Style | Folding, Right-hinged, Auto-close |
For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
Alert:
For indoor use only
Product Code | Brand | Depth (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
15933656
|
asecos™
3023600134387 |
616 mm | 3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£2541.88
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15973696
|
asecos™
3023600234387 |
616 mm | 3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow |
£2541.88
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15973656
|
asecos™
3023600134391 |
616 mm | 3 x tray shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£2743.28
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15913706
|
asecos™
3023600234391 |
616 mm | 3 x tray shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
£2743.28
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15983656
|
asecos™
3023600134392 |
616 mm | 4 x drawer (sheet steel powder-coated) | Light Gray |
£2892.74
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15923706
|
asecos™
3023600234392 |
616 mm | 4 x drawer (sheet steel powder-coated) | Yellow |
£2892.74
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15903666
|
asecos™
3023600134394 |
616 mm | 6 x drawer (sheet steel powder-coated) | Light Gray |
£3172.58
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15943706
|
asecos™
3023600234394 |
616 mm | 6 x drawer (sheet steel powder-coated) | Warning Yellow |
£3172.58
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15974236
|
asecos™
3031600134402 |
749 mm | 4 x drawer (sheet steel powder-coated) | Light Gray |
£3022.06
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15994246
|
asecos™
3031600234402 |
749 mm | 4 x drawer (sheet steel powder-coated) | Warning Yellow |
£3022.06
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15994236
|
asecos™
3031600134404 |
749 mm | 6 x drawer (sheet steel powder-coated) | Light Gray |
£3342.18
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15914256
|
asecos™
3031600234404 |
749 mm | 6 x drawer (sheet steel powder-coated) | Yellow |
£3342.18
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Space saving folding door: less required space in front of the cabinet while door is open
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed: serial automatic door closing (TSA) releases arrested door after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1742 mm | |
Powder-coated plain | |
450 mm |
Steel | |
Folding, Right-hinged, Auto-close | |
1968 mm | |
1 | |
596 mm |