missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Tygon™ E-LFL B/T™ PerfectPostion™ Pump Tubing

Ensure optimal performance from your Masterflex™ B/T series pump

Brand:  Masterflex™ 06449-87

 View more versions of this product

Product Code. 15590652

  • £752.00 / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Nontoxic and nonhemolytic with excellent biocompatibility
  • Long pumping life; pump at continuous pressures up to 40 psi
  • Low extractables and low gas permeability
  • Meets USP Class VI criteria and is FDA compliant
  • High-performance pump tubing for challenging applications
  • Application suitability: Good with acids, alkalis, pressure, vacuum, and excellent with viscous and sterile fluids (Not recommended with Organic solvents)
  • Physical characteristics :
    • PVC-based material with plasticizer.
    • Firm (stiff) material.
    • Transparent, clear.
  • Applications:
    • Laboratory, pharmaceutical processing, and biotech applications for pump lines.
    • Cell and tissue culture work and pharmaceutical uses.
    • Also good for light-sensitive samples.
  • Optimized to provide better performance in higher-pressure applications.
  • Each tubing size is manufactured to extremely close tolerances that match B/T pump heads.
  • These tight tolerances ensure accurate, repeatable flow, and long tubing life.
  • Plus, the PerfectPosition™ tubing retention marks (found on most of the tubing formulations), indicate the best placement of the tubing within the pump head.
Specifications

Specifications

USP Class VI; European Pharmacopoeia (EP 3.2.9); FDA 21 CFR 175.300; EU Food; Reach Compliant (non-DEHP); RoHS Compliant; ADCF Compliant
12.7 mm
25 ft.
0°C
18900 mL/min
EtO, Autoclave
660 mm Hg
Autoclavable
Tygon E-LFL
7.6 m
87
1/2 in.
2.4 bar
40°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers