Learn More
DuPont™ Tychem™ 6000 F with Socks
GREENER_CHOICE
Product Code. 16203512
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Tychem™ 6000 F hooded coveralls with integrated socks that are designed to be worn inside shoes or boots.
  • Made from a lightweight and durable fabric (< 500 g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate
  • Offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards, and certain chemical warfare agents
  • Certified according to Regulation (EU) 2016/425
  • Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
  • EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) on inside
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Self-adhesive double zipper flap closure system for higher protection
  • Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Size 2X-Large
Ankle Style Elastic, Attached socks, Boot overflaps
Closure Type Double zipper with self-adhesive flap and chin flap
Cuff Style Thumb Loop
Description Individually Packed
Disposable Single-use
Gender Unisex
Neck Style Hooded
Quantity 20/Cs.
Seam Type Stitched, Taped
Sterility Non-sterile
Waist Style Elastic