missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Tube Block

Product Code. 15722549
Click to view available options
Holds:
Conical Tubes
Cryotubes
Eppendorf Tubes
Tubes
No. of Places:
4
8
9
24
30
Tube Size:
0.5 mL
1.5 mL
15 mL
2 mL
5 mL
5 to 7 mL
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

OHAUS™ Tube Block is constructed of Valox 357 Plastic and accommodates a variety of tubes.

  • Designed to securely hold tubes in place
  • Applications include: cell cultures, bacterial and yeast cultures, immunoassays, protein studies, blotting techniques, staining and destaining gels. solubility studies, extraction procedures, DNA, RNA, hybridizations, enzyme reactions, ELISA assays
TRUSTED_SUSTAINABILITY

Specifications

Description Block For 24 X 1.5 mL Tubes
For Use With (Equipment) Shaker
Holds Tubes
Material Valox 357 Plastic
No. of Places 24
Type Block
Tube Size 1.5 mL
Dimensions (L x W) 10.20 cm x 14.20 cm
Dimensions (L x W x H) 10.2 x 14.2 x 5.3 cm
Length (English) 4.00 in.
Length (Metric) 10.20 cm
Width (English) 5.60 in.
Width (Metric) 14.20 cm
Height (English) 2.10 in.
Height (Metric) 5.300 cm
Depth (English) 4.00 in.
Depth (Metric) 10.20 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.