missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMT2B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20767-1-AP-20UL
This item is not returnable.
View return policy
Description
Probable S-adenosyl-L-methionine-dependent methyltransferase that catalyzes the formation of 5-methyl-uridine at position 54 (m5U54) in all tRNA. May also have a role in tRNA stabilization or maturation.Specifications
TRMT2B | |
Polyclonal | |
Unconjugated | |
TRMT2B | |
CXorf34, TRM2 homolog, TRMT2B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79979 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96GJ1 | |
TRMT2B | |
TRMT2B Fusion Protein Ag14750 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction