Learn More
Tris SDS PAGE Running Buffer, FASTRun, 10X
Compatible with Tris-Glycine SDS (SDS-PAGE) chemistry whether you buy premade or pour your own (PYO) polyacrylamide gels
Brand: Fisher Bioreagents BP881-1
Code : NEW
Additional Details : CAS Number : 77-86-1 Weight : 1.00000kg
Description
- Cut the time you spend waiting for your SDS PAGE gels to finish by over 70% while providing comparable or better clarity over traditional buffers
- Novel buffer chemistry gives you better resolution, thus requiring fewer TG gel compositions to fully resolve a protein.
Specifications
151-21-3 | |
7.7 to 8.3 (1X/10X soln. at 25°C) | |
1L |
Colorless to Yellow | |
Liquid |