missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS Hydrochloride, Ultrapure Bioreagent, J.T. Baker™
268.85 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Poly Bottle
Poly Pail
Description
- For Liquid Chromatography and Molecular Biology Applications
Specifications
Packaging | Poly Bottle |
Quantity | 500 g |
Formula Weight | 157.6 |
Grade | Ultrapure Bioreagent |
Chemical Name or Material | TRIS Hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'