missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TRIS-buffered saline (TBS, 10X) pH 7.4, for Western blot
Click to view available options
Quantity:
1 L
2 L
4 L
Description
For Western blot washing.TRIS-buffered saline (TBS, 10X) is used as a washing buffer for electrodes, alkaline phosphatase and peroxidase conjugates in dot blot assay. Further, it acts as a pH stabilizer, which enable washing without disruption of antibody-antigen binding interactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
For Western blot washing.TRIS-buffered saline (TBS, 10X) is used as a washing buffer for electrodes, alkaline phosphatase and peroxidase conjugates in dot blot assay. Further, it acts as a pH stabilizer, which enable washing without disruption of antibody-antigen binding interactions.
Solubility
Miscible with water.
Notes
500mM Tris, 1.5M NaCl, pH 7.4.Store in a cool place. Incompatible with strong oxidizing agents.
For Western blot washing.TRIS-buffered saline (TBS, 10X) is used as a washing buffer for electrodes, alkaline phosphatase and peroxidase conjugates in dot blot assay. Further, it acts as a pH stabilizer, which enable washing without disruption of antibody-antigen binding interactions.
Solubility
Miscible with water.
Notes
500mM Tris, 1.5M NaCl, pH 7.4.Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Concentration | 10X |
pH | 7.4 |
Physical Form | Liquid |
Quantity | 1 L |
Solubility Information | Miscible with water. |
Chemical Name or Material | TRIS-buffered saline, for Western blot |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'