missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade

£88.84 - £274.37

Chemical Identifiers

CAS 51805-45-9
Molecular Formula C9H12O6P
Molecular Weight (g/mol) 247.16
MDL Number MFCD00145469
InChI Key PZBFGYYEXUXCOF-UHFFFAOYSA-K
Synonym tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
PubChem CID 2734570
IUPAC Name 3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
SMILES [O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15450127
View Documents Promotion Details
Thermo Scientific Chemicals
J60316.04
2 g
£88.84
2g
Please to purchase this item. Need a web account? Register with us today!
15460127
View Documents
Thermo Scientific Chemicals
J60316.06
5 g
£137.69
5g
Please to purchase this item. Need a web account? Register with us today!
15470127
View Documents Promotion Details
Thermo Scientific Chemicals
J60316.09
10 g
£274.37
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reducing disulfide bonds in peptides and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

51805-45-9
247.16
PZBFGYYEXUXCOF-UHFFFAOYSA-K
2734570
[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O
C9H12O6P
MFCD00145469
tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride

Specifications

51805-45-9
Plastic bottle
MFCD00145469
Moisture sensitive
Soluble in DMSO or water
[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O
247.16
286.65
Molecular Biology
Tris(2-carboxyethyl)phosphine hydrochloride
177°C to 178°C
C9H12O6P
UN3261
tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
PZBFGYYEXUXCOF-UHFFFAOYSA-K
3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
2734570
98%
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'tsca' : No

Recommended Storage : Keep cold

Product Identifier
  • Tris(2-carboxyethyl)phosphine hydrochloride, Molecular Biology Grade

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Tris(2-carboxyethyl)phosphine hydrochloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only