missing translation for 'onlineSavingsMsg'
Learn More

Tris(1-methoxy-2-methyl-2-propoxy)bismuth, 99.99% (metals basis), Thermo Scientific Chemicals

£97.40 - £1437.00

Chemical Identifiers

Molecular Formula C15H33O6Bi
MDL Number MFCD11113030
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15416965
View Documents
Thermo Scientific Alfa Aesar
045488.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15426965
View Documents
Thermo Scientific Alfa Aesar
045488.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15436965
View Documents
Thermo Scientific Alfa Aesar
045488.14
25 g
£1437.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tris(1-methoxy-2-methyl-2-propoxy)bismuth is used as metal organic precursors to prepare bismuth titanium oxide thin films, which finds application in ferroelectric random access memory.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Chemical Identifiers

C15H33O6Bi
MFCD11113030

Specifications

70°C
C15H33O6Bi
Moisture sensitive
518.4
Crystalline
Tris (1-methoxy-2-methyl-2-propoxy)bismuth
Yellow
MFCD11113030
Soluble in ethylcyclohexane.
99.99%
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

missing translation for 'storageNote' :

  • Packaged under Argon

Recommended Storage : Ambient temperatures; Store under Argon

Product Identifier
  • Tris(1-methoxy-2-methyl-2-propoxy)bismuth

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only