Learn More
Tripropargylamine, 97%, Thermo Scientific Chemicals
£36.60 - £560.00
Chemical Identifiers
CAS | 6921-29-5 |
---|---|
Molecular Formula | C9H9N |
Molecular Weight (g/mol) | 131.178 |
MDL Number | MFCD00008577 |
InChI Key | ZHOBJWVNWMQMLF-UHFFFAOYSA-N |
Synonym | tripropargylamine, tri-2-propynylamine, tris propyn-2-yl amine, 2-propyn-1-amine, n,n-di-2-propynyl, unii-5e92gfj1aj, n,n-di-2-propynyl-2-propyn-1-amine, 5e92gfj1aj, n,n-diprop-2-yn-1-ylprop-2-yn-1-amine, tripropargyl amine, acmc-20ajcb |
PubChem CID | 23351 |
IUPAC Name | N,N-bis(prop-2-ynyl)prop-2-yn-1-amine |
SMILES | C#CCN(CC#C)CC#C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15420298
|
Thermo Scientific Alfa Aesar
H53459.03 |
1 g |
£36.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15430298
|
Thermo Scientific Alfa Aesar
H53459.06 |
5 g |
£144.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15440298
|
Thermo Scientific Alfa Aesar
H53459.14 |
25 g |
£560.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tripropargylamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTripropargylamine is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
6921-29-5 | |
131.178 | |
ZHOBJWVNWMQMLF-UHFFFAOYSA-N | |
23351 | |
C#CCN(CC#C)CC#C |
C9H9N | |
MFCD00008577 | |
tripropargylamine, tri-2-propynylamine, tris propyn-2-yl amine, 2-propyn-1-amine, n,n-di-2-propynyl, unii-5e92gfj1aj, n,n-di-2-propynyl-2-propyn-1-amine, 5e92gfj1aj, n,n-diprop-2-yn-1-ylprop-2-yn-1-amine, tripropargyl amine, acmc-20ajcb | |
N,N-bis(prop-2-ynyl)prop-2-yn-1-amine |
Specifications
6921-29-5 | |
79°C to 85°C (11 mmHg) | |
C9H9N | |
1.484 | |
UN1993 | |
ZHOBJWVNWMQMLF-UHFFFAOYSA-N | |
N,N-bis(prop-2-ynyl)prop-2-yn-1-amine | |
23351 | |
97% |
0.927 | |
57°C (135°F) | |
(HC≡CCH2)3N | |
MFCD00008577 | |
tripropargylamine, tri-2-propynylamine, tris propyn-2-yl amine, 2-propyn-1-amine, n,n-di-2-propynyl, unii-5e92gfj1aj, n,n-di-2-propynyl-2-propyn-1-amine, 5e92gfj1aj, n,n-diprop-2-yn-1-ylprop-2-yn-1-amine, tripropargyl amine, acmc-20ajcb | |
C#CCN(CC#C)CC#C | |
131.178 | |
131.18 | |
Tripropargylamine |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 230-038-5
missing translation for 'rtecsNumber' : YK7660000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only