Learn More
Triphosgene, 99%, Thermo Scientific Chemicals
£42.70 - £500.00
Chemical Identifiers
CAS | 32315-10-9 |
---|---|
Molecular Formula | C3Cl6O3 |
Molecular Weight (g/mol) | 296.75 |
InChI Key | UCPYLLCMEDAXFR-UHFFFAOYSA-N |
Synonym | triphosgene, bis trichloromethyl carbonate, methanol, trichloro-, carbonate 2:1, ditrichloromethyl carbonate, triphosgene bis-trichloromethyl carbonate, methanol, 1,1,1-trichloro-, 1,1'-carbonate, triphosgen, tri phosgene, tri-phosgene |
PubChem CID | 94429 |
IUPAC Name | bis(trichloromethyl) carbonate |
SMILES | C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10772471
|
Thermo Scientific Acros
258950050 |
5 g | Glass bottle | N/A | |||||
10513601
|
Thermo Scientific Acros
258950100 |
10 g | Glass bottle | N/A | |||||
10227483
|
Thermo Scientific Acros
258950500 |
50 g | Glass bottle | N/A | |||||
11914701
|
Thermo Scientific Acros
258952500 |
250 g | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent, C-X bond formation reagentChemical Identifiers
32315-10-9 | |
296.75 | |
triphosgene, bis trichloromethyl carbonate, methanol, trichloro-, carbonate 2:1, ditrichloromethyl carbonate, triphosgene bis-trichloromethyl carbonate, methanol, 1,1,1-trichloro-, 1,1'-carbonate, triphosgen, tri phosgene, tri-phosgene | |
bis(trichloromethyl) carbonate |
C3Cl6O3 | |
UCPYLLCMEDAXFR-UHFFFAOYSA-N | |
94429 | |
C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl |
Specifications
32315-10-9 | |
White | |
203.0°C to 206.0°C | |
98.5% min | |
C(Cl3)OC(O)OC(Cl)3 | |
15,9921 | |
triphosgene, bis trichloromethyl carbonate, methanol, trichloro-, carbonate 2:1, ditrichloromethyl carbonate, triphosgene bis-trichloromethyl carbonate, methanol, 1,1,1-trichloro-, 1,1'-carbonate, triphosgen, tri phosgene, tri-phosgene | |
UCPYLLCMEDAXFR-UHFFFAOYSA-N | |
bis(trichloromethyl) carbonate | |
94429 | |
99% | |
Triphosgene |
77.0°C to 81.0°C | |
1.7800g/mL | |
Authentic | |
C3Cl6O3 | |
17,127 | |
1.78 | |
Solubility in water: practically insoluble, may form phosgene. Other solubilities: 588g/L ch2cl2 (20°C), soluble in alcohol, benzene, tetrachloromethane, and ether | |
C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl | |
296.75 | |
296.75 | |
Crystalline Powder, Crystals or Chunks |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 250-986-3
- Danger
Hazard Category
- Acute toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only