missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylsilanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Triphenylsilanol is used as a water surrogate for regioselective Pd catalyzed allylations. A thick on germanium substrates have been produced by the electron bombardment of an evaporated thin film of triphenylsilanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triphenylsilanol is used as a water surrogate for regioselective Pd catalyzed allylations. A thick on germanium substrates have been produced by the electron bombardment of an evaporated thin film of triphenylsilanol.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Triphenylsilanol is used as a water surrogate for regioselective Pd catalyzed allylations. A thick on germanium substrates have been produced by the electron bombardment of an evaporated thin film of triphenylsilanol.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Specifications
Specifications
Melting Point | 149°C to 154°C |
Quantity | 5 g |
Beilstein | 985007 |
Solubility Information | Insoluble in water. |
Formula Weight | 276.41 |
Percent Purity | 98% |
Sensitivity | Moisture sensitive |
Density | 1.17 |
Chemical Name or Material | Triphenylsilanol |
Safety and Handling
Product Identifier
- Triphenylsilanol
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : triphenylsilanol
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'