Learn More
Trimethyltin bromide, Thermo Scientific™
£28.00 - £331.00
Chemical Identifiers
CAS | 1066-44-0 |
---|---|
Molecular Formula | C3H11BrSn |
Molecular Weight (g/mol) | 245.74 |
MDL Number | MFCD00000051 |
InChI Key | KNPKGRQZYPSMBF-UHFFFAOYSA-N |
Synonym | trimethyltin bromide, bromotrimethyltin, stannane, bromotrimethyl, trimethylstannyl bromide, trimethyltin bromide 6ci, me3snbr, tin, bromotrimethyl, acmc-20ajcw, bromo trimethyl stannane, ch3 3snbr |
PubChem CID | 14015 |
IUPAC Name | bromo(trimethyl)stannane |
SMILES | Br.C[SnH](C)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1066-44-0 | |
245.74 | |
KNPKGRQZYPSMBF-UHFFFAOYSA-N | |
14015 | |
Br.C[SnH](C)C |
C3H11BrSn | |
MFCD00000051 | |
trimethyltin bromide, bromotrimethyltin, stannane, bromotrimethyl, trimethylstannyl bromide, trimethyltin bromide 6ci, me3snbr, tin, bromotrimethyl, acmc-20ajcw, bromo trimethyl stannane, ch3 3snbr | |
bromo(trimethyl)stannane |
Specifications
1066-44-0 | |
165°C | |
C3H11BrSn | |
UN2788 | |
trimethyltin bromide, bromotrimethyltin, stannane, bromotrimethyl, trimethylstannyl bromide, trimethyltin bromide 6ci, me3snbr, tin, bromotrimethyl, acmc-20ajcw, bromo trimethyl stannane, ch3 3snbr | |
KNPKGRQZYPSMBF-UHFFFAOYSA-N | |
bromo(trimethyl)stannane | |
14015 | |
Liquid |
27°C | |
>110°C (230°F) | |
MFCD00000051 | |
Moisture sensitive | |
Decomposes in water. | |
Br.C[SnH](C)C | |
245.74 | |
243.7 | |
Trimethyltin bromide |
Safety and Handling
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 213-916-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures