missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | TRIM24 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892734
|
Proteintech
14208-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882734
|
Proteintech
14208-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc binding domains (a RING, a B box type 1 and a B box type 2) and a coiled coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
TRIM24 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
O15164, Q64127 | |
21848, 500084, 8805 | |
TRIM24 Fusion Protein Ag5417 | |
Primary | |
-20°C | |
TRIM24 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
hTIF1, PTC6, RING finger protein 82, RNF82, TF1A, TIF1, TIF1 alpha, TIF1A, TIF1ALPHA, TRIM24, tripartite motif containing 24 | |
Trim24 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |