missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilane, 98%, Thermo Scientific Chemicals

Catalog Number p-205795
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6485-79-6
C9H22Si
158.36
MFCD00009657
YDJXDYKQMRNUSA-UHFFFAOYSA-N
triisopropylsilane, tris propan-2-yl silane, triisopropyl silane, tri propan-2-yl silicon, tri-isopropylsilyl radical, triisopropylsilyl, tri-iso-propylsilane, ambotzrl-1102, pubchem12855
6327611
tri(propan-2-yl)silicon
CC(C)[SiH](C(C)C)C(C)C
This item is not returnable. View return policy
6485-79-6
C9H22Si
158.36
MFCD00009657
YDJXDYKQMRNUSA-UHFFFAOYSA-N
triisopropylsilane, tris propan-2-yl silane, triisopropyl silane, tri propan-2-yl silicon, tri-isopropylsilyl radical, triisopropylsilyl, tri-iso-propylsilane, ambotzrl-1102, pubchem12855
6327611
tri(propan-2-yl)silicon
CC(C)[SiH](C(C)C)C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Density 0.7730g/mL
Boiling Point 84.0°C to 86.0°C (35.0 mmHg)
Flash Point 37°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)2CH]3SiH
Refractive Index 1.4340 to 1.4360
Quantity 10 g
Beilstein 04,IV,3907
Specific Gravity 0.773
Solubility Information Solubility in water: negligible
Formula Weight 158.36
Percent Purity 98%
Chemical Name or Material Triisopropylsilane, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triisopropylsilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'