missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisopropanolamine cyclic borate, 98+%, Thermo Scientific Chemicals
£94.87 - £394.97
Chemical Identifiers
CAS | 101-00-8 |
---|---|
Molecular Formula | C9H18BNO3 |
Molecular Weight (g/mol) | 199.06 |
MDL Number | MFCD03791298 |
InChI Key | IWKGJTDSJPLUCE-UHFFFAOYSA-N |
Synonym | triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate |
PubChem CID | 225550 |
IUPAC Name | 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane |
SMILES | B12OC(CN(CC(O1)C)CC(O2)C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10247500
|
Thermo Scientific Acros
269241000 |
100 g | Glass bottle |
£94.87
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10723275
|
Thermo Scientific Acros
269245000 |
500 g | Glass bottle |
£394.97
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
101-00-8 | |
199.06 | |
IWKGJTDSJPLUCE-UHFFFAOYSA-N | |
225550 | |
B12OC(CN(CC(O1)C)CC(O2)C)C |
C9H18BNO3 | |
MFCD03791298 | |
triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate | |
3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane |
Specifications
101-00-8 | |
White to Yellow | |
98% min. (GC) | |
MFCD03791298 | |
IWKGJTDSJPLUCE-UHFFFAOYSA-N | |
3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane | |
225550 | |
98+% | |
Triisopropanolamine cyclic borate |
150°C to 157°C | |
Authentic | |
C9H18BNO3 | |
triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate | |
B12OC(CN(CC(O1)C)CC(O2)C)C | |
199.06 | |
199.06 | |
Crystalline Chunks and Needles |
Safety and Handling
Product Identifier
- Triisopropanolamine cyclic borate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title