missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexyltin chloride, 97%, Thermo Scientific Chemicals

Product Code. 15471768
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.

Solubility
Soluble in ethanol, ether, toluene and chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3091-32-5
Molecular Formula C18H33ClSn
Molecular Weight (g/mol) 403.622
MDL Number MFCD00003817
InChI Key OJVZYIKTLISAIH-UHFFFAOYSA-M
Synonym tricyclohexyltin chloride, stannane, chlorotricyclohexyl, chlorotricyclohexyltin, chloro tricyclohexyl stannane, tin, chlorotricyclohexyl, tricyclohexyltinchloride, acmc-1ck6z, tricyclohexylchlorotin
PubChem CID 76539
IUPAC Name chloro(tricyclohexyl)stannane
SMILES C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)Cl

Specifications

Melting Point 123°C to 125°C
Linear Formula (C6H11)3SnCl
Quantity 5 g
UN Number UN3146
Solubility Information Soluble in ethanol,ether,toluene and chloroform. Insoluble in water.
Formula Weight 403.6
Percent Purity 97%
Chemical Name or Material Tricyclohexyltin chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'