missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricyclohexyltin chloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.
Solubility
Soluble in ethanol, ether, toluene and chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.
Solubility
Soluble in ethanol, ether, toluene and chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | 123°C to 125°C |
Linear Formula | (C6H11)3SnCl |
Quantity | 1 g |
UN Number | UN3146 |
Solubility Information | Soluble in ethanol,ether,toluene and chloroform. Insoluble in water. |
Formula Weight | 403.6 |
Percent Purity | 97% |
Chemical Name or Material | Tricyclohexyltin chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'