Learn More
Tricyclohexylphosphine, 96%, Thermo Scientific Chemicals
Description
Tricyclohexylphosphine is used as a ligand in organometallic chemistry. As a reagent, it is used for a range of metal-catalyzed organic transformations. Crabtree's catalyst is an organoiridium compound containing tricyclohexyl phosphine used for hydrogenation of mono-, di-, tri-, and tetra- substituted substrates and hydrogen transfer reactions. Grubbs’catalysts consists of transition metal ruthenium and tricylohexyl phosphine utilized in olefin metathesis in synthetic organic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 76°C to 78°C |
Color | White |
Odor | Unpleasant |
Quantity | 1 g |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in most organic solvents. Insoluble in water. |
Formula Weight | 280.42 |
Percent Purity | 96% |
Physical Form | Solid |
Chemical Name or Material | Tricyclohexylphosphine |
RUO – Research Use Only