missing translation for 'onlineSavingsMsg'
Learn More

Tricine, ≥98.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals

Product Code. 15845478
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 5704-04-1
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-Tris(Hydroxymethyl)methylglycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Melting Point ∼183°C (decomposition)
Physical Form Crystalline Powder
Quantity 1 kg
Linear Formula (CH2OH)3CNHCH2COOH
Packaging Plastic bottle
Beilstein 1937804
Formula Weight 179.17
Percent Purity ≥98.5%
Assay Dry basis
Chemical Name or Material Tricine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.