missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Tygon™ E-Lab 3-Stop Stopped Pump Tubing

Designed for optimize performance in your Ismatec™ pumps with this tubing. Tygon™ E-Lab 3-Stop Stopped Pump Tubing is ideal for sensitive pharmaceutical, biotech and general-purpose lab applications.

Marca:  Masterflex™ 96461-42

 vedi altre versioni di questo prodotto

Codice prodotto. 15146516

  • 142.00CHF / pezzi 12
Spedizione stimata: 16-01-2025
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Descrizione

Descrizione

Designed for optimize performance in your Ismatec™ pumps with this tubing. Tygon™ E-Lab 3-Stop Stopped Pump Tubing is ideal for sensitive pharmaceutical, biotech and general-purpose lab applications.

Tubing is resistant to ozone and UV light—won't crack, swell, or harden. Long service life and outstanding fatigue resistance due to flexing. Ultra-smooth inner bore provides excellent biocompatibility. Use this tubing for sterile filling and processing, cell harvest and media process systems, filtration, fermentation, cell and tissue culture transport.
  • Translucent yellow color
  • Temp range: -50 to 74°C
  • Life at 100 rpm: 1000 hrs
  • Heat-sealable, bondable, sterile-weldable and custom moldable for assemblies
  • Non-cytotoxic, non-pyrogenic and non-hemolytic
  • Low protein binding
Specifica

Specifica

Stopped Pump Tubing
Tygon
applications using Acetone, Methyl-ethyl-ketone (MEK), and other highly aggressive chemicals
12 Pack
Translucent Yellow
2.06 mm
40.6 cm
-50°C
0.40 to 59mL/min.
74°C
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali