missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Transferpettor™ Pipetter Station
Provides handy and easily accessible storage for Transferpettor™ instruments and accessories.
Brand: BRAND™ 702890
Code : 02
Additional Details : Weight : 3.67410kg
Description
- Choose to accommodate 2 or 4 instruments up to 200μL
- Holds pipettes upright
- Transferpettors sold separately.
Specifications
Not Autoclavable | |
2 | |
19.05 cm | |
12.7 cm | |
9 in. |
Plastic | |
Transferpettor™ pipetting devices | |
7.5 in. | |
5 in. | |
22.86 cm |