Learn More
trans-2-Octenal, 95%, Thermo Scientific Chemicals
£30.10 - £101.00
Chemical Identifiers
CAS | 2548-87-0 |
---|---|
Molecular Formula | C8H14O |
Molecular Weight (g/mol) | 126.199 |
MDL Number | MFCD00007011 |
InChI Key | LVBXEMGDVWVTGY-VOTSOKGWSA-N |
Synonym | trans-2-octenal, e-oct-2-enal, e-2-octenal, 2-octenal, trans-2-octen-1-al, e-2-octen-1-al, 2-octen-1-al, 2-octenel, 2-octenal, e, 2-octenal, 2eShow More |
PubChem CID | 5283324 |
ChEBI | CHEBI:61748 |
IUPAC Name | (E)-oct-2-enal |
SMILES | CCCCCC=CC=O |
Description
Used in flavoring tangerine, mandarin, cherry, melon and orange and also as a fragrances agent. It causes defects for locomotion in green fluorescent protein (GFP).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in flavoring tangerine, mandarin, cherry, melon and orange and also as a fragrances agent. It causes defects for locomotion in green fluorescent protein (GFP). It is an antimicrobial agent that destroys fungi by suppressing their ability to grow or reproduce.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and fixed oils.
Notes
Air Sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
2548-87-0 | |
126.199 | |
LVBXEMGDVWVTGY-VOTSOKGWSA-N | |
5283324 | |
(E)-oct-2-enal |
C8H14O | |
MFCD00007011 | |
trans-2-octenal, e-oct-2-enal, e-2-octenal, 2-octenal, trans-2-octen-1-al, e-2-octen-1-al, 2-octen-1-al, 2-octenel, 2-octenal, e, 2-octenal, 2eShow More | |
CHEBI:61748 | |
CCCCCC=CC=O |
Specifications
2548-87-0 | |
84°C to 86°C (19 mmHg) | |
Pleasant | |
1.45 | |
Air Sensitive | |
Not miscible or difficult to mix in water. Soluble in alcohol and fixed oils. | |
CCCCCC=CC=O | |
126.199 | |
CHEBI:61748 | |
95% |
0.84 | |
71°C (160°F) | |
C8H14O | |
MFCD00007011 | |
trans-2-octenal, e-oct-2-enal, e-2-octenal, 2-octenal, trans-2-octen-1-al, e-2-octen-1-al, 2-octen-1-al, 2-octenel, 2-octenal, e, 2-octenal, 2e | |
LVBXEMGDVWVTGY-VOTSOKGWSA-N | |
(E)-oct-2-enal | |
5283324 | |
126.2 | |
trans-2-Octenal |
Safety and Handling
GHS H Statement
H315-H317-H227
Causes skin irritation.
May cause an allergic skin reaction.
Combustible liquid.
P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P501c
H227-H315-H317-H319
missing translation for 'einecsNumber' : 219-833-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.