Learn More
trans-1,2-Dibromocyclohexane, 99%, Thermo Scientific Chemicals
£20.30 - £141.00
Chemical Identifiers
CAS | 7429-37-0 |
---|---|
Molecular Formula | C6H10Br2 |
Molecular Weight (g/mol) | 241.95 |
MDL Number | MFCD00003820 |
InChI Key | CZNHKZKWKJNOTE-PHDIDXHHSA-N |
Synonym | trans-1,2-dibromocyclohexane, 1r,2r-1,2-dibromocyclohexane, +/--trans-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, trans-1,2-dibromcyclohexan, 1beta,2alpha-dibromocyclohexane, cyclohexane,2-dibromo-, trans, 1r,2r-1,2-bis bromanyl cyclohexane |
PubChem CID | 98506 |
IUPAC Name | (1R,2R)-1,2-dibromocyclohexane |
SMILES | Br[C@@H]1CCCC[C@H]1Br |
Description
trans-1,2-Dibromocyclohexane is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-1,2-Dibromocyclohexane is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
7429-37-0 | |
241.95 | |
CZNHKZKWKJNOTE-PHDIDXHHSA-N | |
98506 | |
Br[C@@H]1CCCC[C@H]1Br |
C6H10Br2 | |
MFCD00003820 | |
trans-1,2-dibromocyclohexane, 1r,2r-1,2-dibromocyclohexane, +/--trans-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, trans-1,2-dibromcyclohexan, 1beta,2alpha-dibromocyclohexane, cyclohexane,2-dibromo-, trans, 1r,2r-1,2-bis bromanyl cyclohexane | |
(1R,2R)-1,2-dibromocyclohexane |
Specifications
7429-37-0 | |
145°C (100 mmHg) | |
C6H10Br2 | |
MFCD00003820 | |
CZNHKZKWKJNOTE-PHDIDXHHSA-N | |
(1R,2R)-1,2-dibromocyclohexane | |
98506 | |
99% |
1.784 | |
>113°C (235°F) | |
1.5515 | |
trans-1,2-dibromocyclohexane, 1r,2r-1,2-dibromocyclohexane, +/--trans-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, trans-1,2-dibromcyclohexan, 1beta,2alpha-dibromocyclohexane, cyclohexane,2-dibromo-, trans, 1r,2r-1,2-bis bromanyl cyclohexane | |
Br[C@@H]1CCCC[C@H]1Br | |
241.95 | |
241.95 | |
trans-1,2-Dibromocyclohexane |
Safety and Handling
missing translation for 'einecsNumber' : 231-070-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.