missing translation for 'onlineSavingsMsg'
Learn More

TOMM40 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5E2C5, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen TOMM40
Clone 5E2C5
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16828745
View Documents
Proteintech
66658-1-IG-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16818745
View Documents
Proteintech
66658-1-IG-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.
Specifications

Specifications

TOMM40
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
C19orf1, D19S1177E, p38.5, PER EC1, PEREC1, Protein Haymaker, TOM40, TOMM40
Tomm40
IgG1
Protein G
Antibody
5E2C5
Western Blot
Unconjugated
Mouse
Human, Porcine, Mouse, Rat
O96008, Q75Q40, Q9QYA2
102165927, 10452, 308416, 53333
TOMM40 Fusion Protein Ag13065
Primary
-20°C
TOMM40
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.