Learn More
Invitrogen™ TNFRSF6B (DcR3) Human ELISA Kit
Sandwich ELISA Kit
£550.00 - £4280.00
Specifications
Accession Number | O95407 |
---|---|
Assay Sensitivity | 0.3 ng/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15405163
|
Invitrogen™
EHTNFRSF6B |
96 Tests |
£550.00
1 set |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
17894220
|
Invitrogen™
EHTNFRSF6BX10 |
10 x 96 Tests |
£4280.00
Pack of 10 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
17155589
|
Invitrogen™
EHTNFRSF6BX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
The Human TNFRSF6B (DcR3) ELISA quantitates Hu DcR3 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu DcR3. Principle of the method The Human DcR3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors. Several novel members in the TNFR family including DR3, DR4, DR5, and DR6 were recently discovered and function as cell death receptors. Two decoy receptors, DcR1 and DcR2, were recently identified to compete with DR4 and DR5 for their ligand TRAIL binding. A novel decoy receptor was more recently discovered and designated DcR3 and TR6, respectively,. Unlike DcR1 and DcR2, DcR3 is a soluble rather than a membrane associated molecule. DcR3 binds to FasL and LIGHT and inhibits FasL and LIGHT induced apoptosis. DcR3 transcript is expressed in a number of lung and colon carcinomas and in some normal tissues.Specifications
O95407 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
DCR3,DJ583P15.1.1,M68,M68E,TR6 | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
0.3 ng/mL | |
Biotin | |
Other Proteins | |
This ELISA antibody pair detects human DcR3. Other species not determined. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
8771 | |
<12% | |
HRP | |
Apoptosis | |
hCG_2044836, DCR3, DJ583P15.1.1, M68, M68E, TR6 | |
1 hr. 20 min. |