Learn More
TNF alpha Monoclonal Antibody (TNF-D), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience BMS154
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: BMS154 (TNF-D) specifically reacts with natural human TNF-alpha as well as with recombinant human TNF-alpha. BMS154 selectively neutralizes human TNF-alpha bioactivity. BMS154 is suitable as primary antibody in staining for FACS analysis. TNF-D is suitable as capture or detection antibody in sandwich ELISA but is not suitable for Western blot analysis. Applications Tested: ELISA, Flow Cytometry, Functional Studies (Blocking). Purity: >90%.
TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.Specifications
TNF alpha | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P01375 | |
Tnf | |
Rec. Human TNF alpha | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Flow Cytometry, Functional Assay | |
TNF-D | |
Unconjugated | |
Tnf | |
APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF a; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFa; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor | |
Mouse | |
Affinity chromatography | |
RUO | |
7124 | |
4° C | |
Liquid |
For Research Use Only.