missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TN buffer soln., Thermo Scientific™
£23.31 - £34.86
Specifications
Chemical Name or Material | TN buffer soln. |
---|---|
Color | Colorless |
Physical Form | Liquid |
Synonym | TRIS NaCl buffer soln |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
TN buffer soln. | |
Colorless | |
Liquid | |
TRIS NaCl buffer soln |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title