missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM35 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24786-1-AP-150UL
This item is not returnable.
View return policy
Specifications
TMEM35 | |
Polyclonal | |
Unconjugated | |
TMEM35A | |
TMEM35, transmembrane protein 35 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
59353, 67564 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q53FP2, Q9D328 | |
TMEM35A | |
TMEM35 Fusion Protein Ag20589 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |