Learn More
Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, Blue
Handle small biopsies and fragile specimens with Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, the ultimate cassettes for specimen security.
Brand:  Epredia™ C-1000-BL
Code : 50
Additional Details : Weight : 1.55700kg
 View more versions of this product
Product Code. 12658346
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Saves time, offers the greatest specimen security, and provides ease of use
  • Provides an inner chamber lined with a “see-through" mesh for easy identification of specimens
  • Eliminates the need for messy sponges or lens paper
  • Ensures tissues are safely processed as cassettes will not pop open during processing
Specifications
Specifications
Mesh Pore Insert
Blue
HistoScreen™ Cassette