missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) acetate, Thermo Scientific Chemicals

£167.00 - £609.00

Chemical Identifiers

CAS 2800-96-6
Molecular Formula C8H12O8Sn
Molecular Weight (g/mol) 354.886
MDL Number MFCD00014976
InChI Key YJGJRYWNNHUESM-UHFFFAOYSA-J
Synonym tin iv acetate, tin acetate, tetraacetoxystannane, tin-iv-acetate, acmc-20ajhv, tin 4+ ion tetraacetate, tetraacetoxytin, tin 4+ tetraacetate
PubChem CID 9863446
IUPAC Name tin(4+);tetraacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sn+4]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11473860
View Documents
Thermo Scientific Alfa Aesar
089921.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11483860
View Documents
Thermo Scientific Alfa Aesar
089921.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Tin(IV) acetate is used to prepare tin-based oxides (Sn-Zn-P-O), which is a negative electrodes for rechargeable lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2800-96-6
354.886
YJGJRYWNNHUESM-UHFFFAOYSA-J
9863446
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sn+4]
C8H12O8Sn
MFCD00014976
tin iv acetate, tin acetate, tetraacetoxystannane, tin-iv-acetate, acmc-20ajhv, tin 4+ ion tetraacetate, tetraacetoxytin, tin 4+ tetraacetate
tin(4+);tetraacetate

Specifications

232°C to 233°C
Crystalline
MFCD00014976
Soluble in acetone and benzene.
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sn+4]
354.886
354.89
Tin(IV) Acetate
2800-96-6
C8H12O8Sn
tin iv acetate, tin acetate, tetraacetoxystannane, tin-iv-acetate, acmc-20ajhv, tin 4+ ion tetraacetate, tetraacetoxytin, tin 4+ tetraacetate
YJGJRYWNNHUESM-UHFFFAOYSA-J
tin(4+);tetraacetate
9863446
Hygroscopic
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only