missing translation for 'onlineSavingsMsg'
Learn More

Tin(Ii) Chloride, Honeywell

Product Code. 15681830
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
PubChem CID 24479
ChEBI CHEBI:78067
IUPAC Name dichlorotin
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Quantity 2 kg
Linear Formula SnCl2
UN Number 3260
Formula Weight 189.62g/mol
Percent Purity 98%
Grade Reagent
Chemical Name or Material Tin(Ii) Chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'