missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) 2-ethylhexanoate, 95%, Thermo Scientific Chemicals

£26.43 - £53.67

Chemical Identifiers

CAS 301-10-0
Molecular Formula C16H30O4Sn
Molecular Weight (g/mol) 405.122
MDL Number MFCD00002676
InChI Key KSBAEPSJVUENNK-UHFFFAOYSA-L
Synonym stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
PubChem CID 9318
IUPAC Name 2-ethylhexanoate;tin(2+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11475637
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23612.14
25 g
£26.43
25g
Please to purchase this item. Need a web account? Register with us today!
11485637
View Documents
Thermo Scientific Alfa Aesar
B23612.22
100 g
£53.67
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, a lubricant, addition agent and a stabilizer for transformer oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, a lubricant, addition agent and a stabilizer for transformer oils.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

301-10-0
405.122
KSBAEPSJVUENNK-UHFFFAOYSA-L
9318
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
C16H30O4Sn
MFCD00002676
stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
2-ethylhexanoate;tin(2+)

Specifica

301-10-0
>110°C (230°F)
C16H30O4Sn
MFCD00002676
Miscible with water.
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
405.122
405.11
Tin(II) 2-ethylhexanoate
1.251
Characteristic
1.494
stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
KSBAEPSJVUENNK-UHFFFAOYSA-L
2-ethylhexanoate;tin(2+)
9318
96%
Videos
Sicurezza e movimentazione

Sicurezza e movimentazione

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c

H317-H318-H361

missing translation for 'einecsNumber' : 206-108-6

missing translation for 'rtecsNumber' : MO7870000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Identificativo prodotto
  • Tin(II) 2-ethylhexanoate

Avvertenza
  • Danger

Categoria di pericolo
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1

Indicazioni di pericolo
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.

Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Informazioni supplementari
  • MIXTURE LIST-Contains : Tin(II) 2-ethylhexanoate

SDS
Documenti

Documenti

Certificati

RUO – Research Use Only