missing translation for 'onlineSavingsMsg'
Learn More

THOC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen THOC1
Concentration 0.33 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16807633
View Documents
Proteintech
10920-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16897623
View Documents
Proteintech
10920-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody recognizes the 84 kDa THOC1 protein.

TCEB1 is known as protein elongin C, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. Western blots using two different antibodies (P100962-T200 and P100963_P050 / P100963_P100) against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
Specifications

Specifications

THOC1
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Rabbit
Mouse, Rat, Human
P59924, Q8R3N6, Q96FV9
225160, 291797, 9984
THOC1 Fusion Protein Ag1352
Primary
-20°C
THOC1
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
HPR1, hTREX84, Nuclear matrix protein p84, P84, P84N5, THO complex 1, THO complex subunit 1, Tho1, THOC1
Thoc1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications