missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Thermo Scientific™ Nalgene™ Stainless Steel Autoclavable Dolly
For moving smalls tanks and carboys
Brand: Thermo Scientific™ 2624-0020
Code : NEW
Additional Details : Weight : 8.35000kg
Description
- Noncorrosive, made of stainless steel
- Casters do not leave marks on floor
Specifications
20.9 in. | |
To move small tanks and carboys | |
227.3 kg |
53 cm | |
Round | |
Stainless Steel |