missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil™ Green ENV™ Environmental Analysis HPLC Column, 5 μm, 4.6 mm x 250 mm

Carbon Load: 7%; Pore Size: 120 Å

Brand:  Thermo Scientific™ 31205-254630

Product Code. 11532901

  • £783.90 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

4.6 mm
5 μm
L1
Environmental Analysis
250 mm
120 Å
7%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.