missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 1-Step CHO High-Yield IVT Kit

Express of functional proteins with much greater yield per mL of reaction than rabbit reticulocyte lysate-based mammalian IVT systems using the Thermo Scientific™ 1-Step CHO High-Yield In Vitro Translation (IVT) Kit.

£244.55 - £1555.84

Specifications

Formulation Multi-component kit
Type 1-step CHO High-Yield IVT Kit
Content And Storage Store in ultra-cold freezer (-68°C to -85°C)
View More Specs

Products 2
Product Code Brand Sufficient For Price Quantity & Availability  
Product Code Brand Sufficient For Price Quantity & Availability  
15302617
View Documents
Thermo Scientific™
88894
2 reactions of 2 mL each This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15392607
View Documents
Thermo Scientific™
88893
2 Reactions of 100μL each This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

The Thermo Scientific 1-Step CHO High-Yield In Vitro Translation (IVT) Kit enables the expression of functional proteins with up to 750 times greater yield per mL of reaction than rabbit reticulocyte lysate-based mammalian IVT systems that only yield up to 10μg protein/mL reaction.

  • High expression - up to 750μg/mL of expressed protein
  • Reproducible results - low variability between experiments
  • Fast - express high levels of protein with 6 hours to overnight incubation
  • Easier storage - allows you to store lyophilized CHO cell lysate at -20°C
  • Functional - obtain functionally active proteins, including those containing disulfide bonds
  • Easy-to-use - three reaction components together carry out transcription and translation in one step
  • Great for labeling - amenable to incorporation of labeled and unnatural amino acids
  • Vector compatibility - uses existing pT7CFE1 vectors for protein expression
The 1-Step CHO High-Yield IVT System uses extracts containing the robust translation machinery of the pharmaceutical industry standard Chinese Hamster Ovary (CHO) cell line to generate functional full-length proteins. In this system, protein expression is performed in a proprietary dialysis device that allows a continuous supply of nucleotides, amino acids, and energy-generating substrates into the reaction while removing inhibitors of proteins synthesis. This continuous-exchange cell-free (CECF) system enables protein expression in an overnight incubation of up to 750μg/mL. The CHO IVT kit includes all of the components required for transcription and translation of a recombinant gene, including an optimized expression vector.
Includes:
Lyophilized cell lysate, reaction mix, accessory proteins, 4X dialysis buffer, control DNA, cloning vector, dialysis devices, and centrifuge tubes.
Requires:
30°C incubator or water bath (shaker incubator increases expression by 30 to 50%)
Applications:
  • Production of large amounts of recombinant protein for structural analysis
  • Generation of active proteins for downstream applications such as protein interaction studies
  • Rapid large-scale expression and characterization of mutant proteins
  • Production of viral particles
  • Incorporation of unnatural amino acids
  • Production of cytotoxic proteins

Specifications

Specifications

Multi-component kit
1-step CHO High-Yield IVT Kit
Store in ultra-cold freezer (-68°C to -85°C)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.