missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Texas Red™-X, Succinimidyl Ester, single isomer
Texas Red™-X, Succinimidyl Ester, single isomer
Brand: Molecular Probes™ T20175
Code : 50
Additional Details : Weight : 0.00500kg
Description
The amine-reactive Texas Red™-X, succinimidyl ester can be used to can be used to create bright red-fluorescent bioconjugates with excitation/emission maxima ∼595/615 nm. This reactive dye contains an additional seven-atom aminohexanoyl spacer ('X') between the fluorophore and the succinimidyl ester group. This spacer helps to separate the fluorophore from its point of attachment, potentially reducing the interaction of the fluorophore with the biomolecule to which it is conjugated.Specifications
2 mg | |
Store in freezer -5°C to -30°C and protect from light. | |
Room Temperature | |
Texas Red™ | |
Texas Red™ | |
595 |
Succinimidyl Ester | |
Amine | |
Active Ester, Succinimidyl Ester | |
Classic Dyes | |
615 |
For Research Use Only. Not for use in diagnostic procedures.