Learn More
Molecular Probes™ Texas Red™ C2-Dichlorotriazine
Texas Red™ C2-dichlorotriazine
Brand: Molecular Probes™ T30200
This item is not returnable.
View return policy
Description
Texas Red C2-dichlorotriazine is a reactive dye with absorption/emission maxima of ∼588/601 nm. Dichlorotriazines readily modify amines in proteins, and are among the few reactive groups that are reported to react directly with polysaccharides and other alcohols in aqueous solution, provided that the pH is >9 and that other nucleophiles are absent.Specifications
5 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Texas Red™ | |
Texas Red™ |
C2-Dichlorotriazine | |
Alcohol, Amine | |
Dichlorotriazine | |
Classic Dyes | |
601 |
For Research Use Only. Not for use in diagnostic procedures.