Learn More
Tetramethylsilane, 99.9+%, NMR grade, Thermo Scientific Chemicals
£30.05 - £73.26
Chemical Identifiers
CAS | 75-76-3 |
---|---|
Molecular Formula | C4H12Si |
Molecular Weight (g/mol) | 88.23 |
MDL Number | MFCD00008274 |
InChI Key | CZDYPVPMEAXLPK-UHFFFAOYSA-N |
Synonym | silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073 |
PubChem CID | 6396 |
ChEBI | CHEBI:85361 |
IUPAC Name | tetramethylsilane |
SMILES | C[Si](C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10615192
|
Thermo Scientific Acros
138470250 |
25 g | Glass bottle |
£30.05
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10553301
|
Thermo Scientific Acros
138471000 |
100 g | Glass bottle |
£73.26
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
75-76-3 | |
88.23 | |
CZDYPVPMEAXLPK-UHFFFAOYSA-N | |
6396 | |
tetramethylsilane |
C4H12Si | |
MFCD00008274 | |
silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073 | |
CHEBI:85361 | |
C[Si](C)(C)C |
Specifications
75-76-3 | |
Colorless | |
26.0°C to 28.0°C | |
99.9% min. (GC) | |
Si(CH3)4 | |
MFCD00008274 | |
15, 9377 | |
silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073 | |
CZDYPVPMEAXLPK-UHFFFAOYSA-N | |
tetramethylsilane | |
6396 | |
88.22 | |
Volatile Liquid |
-99.0°C | |
0.6400g/mL | |
−27°C | |
C4H12Si | |
1.3585 | |
04, 625 | |
0.64 | |
Solubility in water: 0.02g/L (25°C). Other solubilities: soluble in most common organic solvents | |
C[Si](C)(C)C | |
88.23 | |
CHEBI:85361 | |
99.9+% | |
Tetramethylsilane, 99.9+% |
Safety and Handling
GHS H Statement:
Extremely flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-899-1
missing translation for 'tsca' : TSCA
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 1
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only