Learn More
Tetramethylgermanium, 98%, Thermo Scientific Chemicals
£145.00 - £568.00
Chemical Identifiers
CAS | 865-52-1 |
---|---|
Molecular Formula | C4H12Ge |
Molecular Weight (g/mol) | 132.77 |
MDL Number | MFCD00014843 |
InChI Key | ZRLCXMPFXYVHGS-UHFFFAOYSA-N |
Synonym | tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference |
PubChem CID | 70079 |
IUPAC Name | tetramethylgermane |
SMILES | C[Ge](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488756
|
Thermo Scientific Alfa Aesar
A16931.06 |
5 g |
£145.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498756
|
Thermo Scientific Alfa Aesar
A16931.14 |
25 g |
£568.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Employed as a Chemical Vapor Deposition (CVD) precursor to germanium oxide thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as a Chemical Vapor Deposition (CVD) precursor to germanium oxide thin films.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Chemical Identifiers
865-52-1 | |
132.77 | |
ZRLCXMPFXYVHGS-UHFFFAOYSA-N | |
70079 | |
C[Ge](C)(C)C |
C4H12Ge | |
MFCD00014843 | |
tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference | |
tetramethylgermane |
Specifications
865-52-1 | |
0.975 | |
−37°C (−35°F) | |
1.389 | |
UN1993 | |
Not miscible or difficult to mix in water. | |
C[Ge](C)(C)C | |
132.77 | |
132.73 | |
Tetramethylgermanium |
-88°C | |
42°C to 44°C | |
C4H12Ge | |
MFCD00014843 | |
tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference | |
ZRLCXMPFXYVHGS-UHFFFAOYSA-N | |
tetramethylgermane | |
70079 | |
98% |
Safety and Handling
GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c
H224-H302+H312+H332-H319
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: I
missing translation for 'einecsNumber' : 212-745-0
missing translation for 'rtecsNumber' : LY5345000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Tetramethylgermanium
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 1
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : Tetramethylgermanium
RUO – Research Use Only