missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals

£41.71 - £534.66

Chemical Identifiers

CAS 109-99-9, 128-37-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10674672
View Documents
Thermo Scientific Acros
176630010
1 L Glass Bottle
£41.71
1 L
Please to purchase this item. Need a web account? Register with us today!
10499640
View Documents Promotion Details
Thermo Scientific Acros
176630025
2.5 L Glass Bottle
£80.20
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10538143
View Documents
Thermo Scientific Acros
176630100
10 L Plastic Drum
£252.26
10 L
Please to purchase this item. Need a web account? Register with us today!
10552201
View Documents
Thermo Scientific Acros
176630250
25 L Metal Drum
£534.66
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-99-9, 128-37-0
72.11
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
C4H8O
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
CHEBI:26911
C1CCOC1

Specifications

-108.4°C
0.8800g/mL
−21°C
1.407
0.005% max.
C4H8O
0.55 mPa.s (20°C)
0.05% max. (Coulometric)
15, 9356
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
C1CCOC1
72.11
CHEBI:26911
≥99%
Tetrahydrofuran
109-99-9, 128-37-0
66.0°C
Liquid
99+%
0.88
MFCD00005356
17, 10
01,1140; 02,398; 03,278; 05,649; 06,570
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
Extra Pure
Videos
Safety and Handling

Safety and Handling

GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed.

Danger

Product Identifier
  • Tetrahydrofuran

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.