missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachloroethylene, Certified AR for Analysis, Fisher Chemical™
£259.00 - £859.00
Chemical Identifiers
CAS | 127-18-4 |
---|---|
Molecular Formula | C2Cl4 |
Molecular Weight (g/mol) | 165.82 |
MDL Number | MFCD00000834 |
InChI Key | CYTYCFOTNPOANT-UHFFFAOYSA-N |
Synonym | tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin |
PubChem CID | 31373 |
ChEBI | CHEBI:17300 |
IUPAC Name | 1,1,2,2-tetrachloroethene |
SMILES | ClC(Cl)=C(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10569390
|
Fisher Chemical
T/0601/17 |
2.5 L | Amber glass bottle |
£305.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10192300
|
Fisher Chemical
T/0601/PB17 |
2.5 L | Plastic Coated Glass bottle |
£259.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12658655
|
Fisher Chemical
T/0601/25 |
25 L | Metal can |
£859.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
127-18-4 | |
165.82 | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
31373 | |
1,1,2,2-tetrachloroethene |
C2Cl4 | |
MFCD00000834 | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
CHEBI:17300 | |
ClC(Cl)=C(Cl)Cl |
Specifications
127-18-4 | |
Colorless | |
C2Cl4 | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
ClC(Cl)=C(Cl)Cl | |
165.82 | |
CHEBI:17300 | |
18mbar at 20°C |
-22°C | |
120°C | |
MFCD00000834 | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
1,1,2,2-tetrachloroethene | |
31373 | |
165.83g/mol | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Tetrachloroethylene
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/A§/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title