missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachloroethylene, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
£225.00 - £876.00
Chemical Identifiers
CAS | 127-18-4 |
---|---|
Molecular Formula | C2Cl4 |
Molecular Weight (g/mol) | 165.82 |
MDL Number | MFCD00000834 |
InChI Key | CYTYCFOTNPOANT-UHFFFAOYSA-N |
Synonym | tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin |
PubChem CID | 31373 |
ChEBI | CHEBI:17300 |
IUPAC Name | 1,1,2,2-tetrachloroethene |
SMILES | ClC(Cl)=C(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15508624
|
Avantor J.T.Baker
9465-01 |
500 mL | Narrow Mouth Amber Glass Bottle |
£225.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15518624
|
Avantor J.T.Baker
9465-03 |
4 L | Narrow Mouth Amber Glass Bottle |
£876.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
127-18-4 | |
165.82 | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
31373 | |
1,1,2,2-tetrachloroethene |
C2Cl4 | |
MFCD00000834 | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
CHEBI:17300 | |
ClC(Cl)=C(Cl)Cl |
Specifications
127-18-4 | |
MFCD00000834 | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
ClC(Cl)=C(Cl)Cl | |
165.82 | |
CHEBI:17300 | |
BAKER ANALYZED™ Reagent |
C2Cl4 | |
1L = 1.62kg | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
1,1,2,2-tetrachloroethene | |
31373 | |
165.83 | |
Tetrachloroethylene |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
Hazard Statement
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P281-Use personal protective equipment as required.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P391-Collect spillage.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title